EMAIL THIS PAGE TO A FRIEND
Applied catalysis. A, General
Reactions of Acenaphthenequinone and Aceanthrenequinone with Arenes in Superacid.
PMID 21941409
Abstract
The hydroxyalkylation reactions of aceanthrenequinone (6) and acenapthenequinone (7) with a series of arenes have been studied. In reactions with the Brønsted superacid CF(3)SO(3)H (triflic acid), the condensation products are formed in good yields (58-99%, 10 examples) with high regioselectivity. Computational studies were also done to examine the structures and energies of mono- and diprotonated species from 6 and 7. The results from the condensation reactions are consistent with the formation of superelectrophilic species involving protosolvation of carboxonium ion intermediates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
216690
2,2,2-Triphenylacetophenone, 98%
C26H20O
327972
Aceanthrenequinone, 96%
C16H8O2