EMAIL THIS PAGE TO A FRIEND
International journal of food sciences and nutrition
The inhibiting activity of areca inflorescence extracts on human low density lipoprotein oxidation induced by cupric ion.
PMID 21942744
Abstract
The oxidative modification of human low density lipoprotein (LDL) plays a significant role in atherosclerosis. In this study, the inhibiting activity of areca inflorescence extracts (AIEs) on LDL oxidation was investigated by an in vitro study with Trolox as the standard antioxidant. The kinetics of LDL oxidation, thiobarbituric acid reactive substances assay, ferric-reducing antioxidant power assay and copper chelation assay were also evaluated to assess the antioxidant activities of AIEs, and the results revealed that AIEs could delay the lag time and inhibit the formation of malondialdehyde in the process of LDL peroxidation induced by Cu(2+). The boiled water extract displayed the highest antioxidant activity compared with the ambient water extract and ethanol extract. The total phenolic contents and phenolic components of AIEs were also measured by high performance liquid chromatography method. Epicatechin, gallic acid and coumalic acid were the primary phenolic acids in AIEs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C85409
Coumalic acid, 97%
C6H4O4