EMAIL THIS PAGE TO A FRIEND
Chemistry & biology
A systematic family-wide investigation reveals that ~30% of mammalian PDZ domains engage in PDZ-PDZ interactions.
PMID 21944753
Abstract
PDZ domains are independently folded modules that typically mediate protein-protein interactions by binding to the C termini of their target proteins. However, in a few instances, PDZ domains have been reported to dimerize with other PDZ domains. To investigate this noncanonical-binding mode further, we used protein microarrays comprising virtually every mouse PDZ domain to systematically query all possible PDZ-PDZ pairs. We then used fluorescence polarization to retest and quantify interactions and coaffinity purification to test biophysically validated interactions in the context of their full-length proteins. Overall, we discovered 37 PDZ-PDZ interactions involving 46 PDZ domains (~30% of all PDZ domains tested), revealing that dimerization is a more frequently used binding mode than was previously appreciated. This suggests that many PDZ domains evolved to form multiprotein complexes by simultaneously interacting with more than one ligand.