EMAIL THIS PAGE TO A FRIEND
Dental materials journal
Hydroxyapatite particle characteristics influence the enhancement of the mechanical and chemical properties of conventional restorative glass ionomer cement.
PMID 21946488
Abstract
The aims of this study were to improve the mechanical and chemical properties of conventional restorative glass ionomer cement (GIC) by adding hydroxyapatite (HAp) preparations with different characteristics, and to investigate the underlying reaction mechanisms. Fuji IX GP® was used as the control GIC. The experimental GICs consisted of four HAp-particles with different characteristics added at 8 mass% to Fuji IX-powder. All cements were prepared by mixing with Fuji IX-liquid (P/L=3.6). Four HAp-particles were analyzed, and then the mechanical strengths and the fluoride-ion- release-recharge-behaviors of five GIC groups were evaluated. The results of this study demonstrate that the addition of HAp particles with highly reactive properties such as high specific surface area can enhance the flexural strength and fluoride ion release properties of conventional restorative GIC. Our results further indicate that HAp functions as an adsorbent and an ion exchangeable agent, resulting in improved mechanical and chemical properties of GIC.