EMAIL THIS PAGE TO A FRIEND
The EMBO journal
Structural basis for cell surface patterning through NetrinG-NGL interactions.
PMID 21946559
Abstract
Brain wiring depends on cells making highly localized and selective connections through surface protein-protein interactions, including those between NetrinGs and NetrinG ligands (NGLs). The NetrinGs are members of the structurally uncharacterized netrin family. We present a comprehensive crystallographic analysis comprising NetrinG1-NGL1 and NetrinG2-NGL2 complexes, unliganded NetrinG2 and NGL3. Cognate NetrinG-NGL interactions depend on three specificity-conferring NetrinG loops, clasped tightly by matching NGL surfaces. We engineered these NGL surfaces to implant custom-made affinities for NetrinG1 and NetrinG2. In a cellular patterning assay, we demonstrate that NetrinG-binding selectivity can direct the sorting of a mixed population of NGLs into discrete cell surface subdomains. These results provide a molecular model for selectivity-based patterning in a neuronal recognition system, dysregulation of which is associated with severe neuropsychological disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N4037 Anti-Netrin-G1a antibody produced in goat, affinity isolated antibody, lyophilized powder
D1145 Dulbecco’s Modified Eagle’s Medium - high glucose, With 4500 mg/L glucose and sodium bicarbonate, without L-glutamine, sodium pyruvate, and phenol red, liquid, sterile-filtered, suitable for cell culture
D6171 Dulbecco’s Modified Eagle’s Medium - high glucose, HEPES modification, With 4500 mg/L glucose, 25 mM HEPES, and sodium bicarbonate, without L-glutamine and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
D0422 Dulbecco’s Modified Eagle’s Medium - high glucose, With 4500 mg/L glucose and sodium bicarbonate, without L-methionine, L-cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
D5648 Dulbecco’s Modified Eagle’s Medium - high glucose, With 4500 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
D7777 Dulbecco’s Modified Eagle’s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium pyruvate, without sodium bicarbonate, powder, suitable for cell culture
D1152 Dulbecco’s Modified Eagle’s Medium - high glucose, HEPES Modification, With 4500 mg/L glucose, L-glutamine, and 25 mM HEPES, without sodium bicarbonate and pyruvate, powder, suitable for cell culture
D0819 Dulbecco’s Modified Eagle’s Medium - high glucose, AQmedia, With 4500 mg/L glucose, L-alanyl-glutamine, and sodium bicarbonate, without sodium pyruvate., liquid, sterile-filtered, suitable for cell culture
D0822 Dulbecco’s Modified Eagle’s Medium - high glucose, AQmedia, With 4500 mg/L glucose, L-Ala-L-Gln, sodium pyruvate and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture