EMAIL THIS PAGE TO A FRIEND
Journal of applied microbiology
Response of Lactobacillus casei BL23 to phenolic compounds.
PMID 21951613
Abstract
To determine the inhibitory effect of phenolic compounds on Lactobacillus casei BL23, the role of two component signal transduction systems (TCS) and the response of Lact. casei BL23 to p-coumaric acid. Growth of Lact. casei BL23 and 17 derivative strains defective in each TCS harboured by this strain in the presence of p-coumaric acid, ferulic acid, caffeic acid or methyl gallate was monitored. Furthermore, changes in the protein content of Lact. casei BL23 when exposed to p-coumaric acid were evaluated by 2D-SDS-PAGE. Eleven proteins differentially expressed in the presence of p-coumaric acid were detected. Six of them could be identified: ClpP and HtrA, involved in protein turnover and folding, acetyl-CoA carboxylase, involved in lipid metabolism, and an arginyl-tRNA synthetase were more abundant, whereas PurL and PurN, involved in purine biosynthesis, were less abundant. No significant differences were observed between the parental strain and the TCS-defective mutants. p-Coumaric acid elicited a response against membrane and cytoplasmic damages. The inhibitory effect of phenolic compounds on Lact. casei BL23 has been determined. For the first time, cytoplasmic proteins presumably involved in the response of Lact. casei BL23 against p-coumaric acid have been identified.