EMAIL THIS PAGE TO A FRIEND
Acta haematologica
The oral iron chelator deferasirox induces apoptosis in myeloid leukemia cells by targeting caspase.
PMID 21951998
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000537 Emedastine difumarate, European Pharmacopoeia (EP) Reference Standard
C25H34N4O9