EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Fluorescent coronene monoimide gels via H-bonding induced frustrated dipolar assembly.
PMID 21952511
Abstract
Two stage self-assembly of novel coronene monoimide (CMI) based gels that results in resurfacing of monomer emission in the aggregated state is reported. This process is attributed to a frustrated head-head dipolar assembly forced by hydrogen bonding.