EMAIL THIS PAGE TO A FRIEND
Journal of the American Society for Mass Spectrometry
Infrared multiple photon dissociation action spectroscopy and theoretical studies of triethyl phosphate complexes: effects of protonation and sodium cationization on structure.
PMID 21952899
Abstract
The gas-phase structures of protonated and sodium cationized complexes of triethyl phosphate, [TEP + H](+) and [TEP + Na](+), are examined via infrared multiple photon dissociation (IRMPD) action spectroscopy using tunable IR radiation generated by a free electron laser, a Fourier transform ion cyclotron resonance mass spectrometer with an electrospray ionization source, and theoretical electronic structure calculations. Measured IRMPD action spectra are compared to linear IR spectra calculated at the B3LYP/6-31 G(d,p) level of theory to identify the structures accessed in the experimental studies. For comparison, theoretical studies of neutral TEP are also performed. Sodium cationization and protonation produce changes in the central phosphate geometry, including an increase in the alkoxy ∠OPO bond angle and shortening of the alkoxy P-O bond. Changes associated with protonation are more pronounced than those produced by sodium cationization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
538728
Triethyl phosphate, ReagentPlus®, ≥99.8%
C6H15O4P