EMAIL THIS PAGE TO A FRIEND
Synapse
Running wheel exercise ameliorates methamphetamine-induced damage to dopamine and serotonin terminals.
PMID 21953518
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB152
Anti-Tyrosine Hydroxylase Antibody, clone, Chemicon®, from rabbit