EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Ion-beam-induced desorption as a method for probing the stability of the molecule-substrate interface in self-assembled monolayers.
PMID 21954003