EMAIL THIS PAGE TO A FRIEND
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals
Comparison of the course of biomarker changes and kidney injury in a rat model of drug-induced acute kidney injury.
PMID 21955166
Abstract
To aid in evaluating the performance of biomarkers, we measured kidney injury biomarkers in rat models of drug-induced acute kidney injury. Rats were treated with site-specific nephrotoxins, puromycin, gentamicin, cisplatin, or 2-bromoethylamine. Fifteen biomarkers (β-2-microglobulin, calbindin, clusterin, cystatin-C, KIM-1, GST-α, GST-μ, NGAL, osteopontin, EGF, TIMP-1, VEGF, albumin, RPA-1, and urinary total protein) were examined in comparison with BUN, serum creatinine, and NAG. Some biomarkers, which were different depending in each nephrotoxin, showed ability to detect the prodromal stage of drug-induced kidney injury. Characteristic changing patterns of biomarkers were also found depending on the specific lesion site in the kidney. These data suggested that establishment of a suitable biomarker panel would facilitate detection of site-specific kidney injury with high sensitivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06670
2-Bromoethylamine hydrobromide, purum, ≥97.0% (AT)
C2H6BrN · HBr
B65705
2-Bromoethylamine hydrobromide, 99%
C2H6BrN · HBr