EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Tailored materials for high-performance MgB(2) wire.
PMID 21956864
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
553913
Magnesium boride, powder, −100 mesh, ≥99% trace metals basis
B2Mg