EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Enzymatic synthesis of fatty acid ethyl esters by utilizing camellia oil soapstocks and diethyl carbonate.
PMID 21958524
Abstract
This study was reported on a novel process for fatty acid ethyl esters preparation by transesterification and esterification from renewable low-cost feedstock camellia oil soapstocks and friendly acyl acceptor diethyl carbonate. The main components of product were 83.9% ethyl oleate, 8.9% ethyl palmitate, 4.7% ethyl linoleate and 2.1% ethyl stearate, which could be used as eco-friendly renewable resources or additives of industrial solvent and fossil fuel. The effects of molar ratio of diethyl carbonate to soapstocks oil, lipases, organic solvent, reaction temperature and time were investigated, and process conditions were optimized. The yield was up to 98.4% in solvent-free system with molar ratio of diethyl carbonate to soapstocks oil 3:1 and 5% Novozym 435 (based on the weight of soapstocks oil) at 50 °C and 180 rpm for 24 h. Moreover, there was no obvious loss in the yield after lipases were reused for 10 batches without treatment under optimized conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
517135
Diethyl carbonate, anhydrous, ≥99%
C5H10O3
D91551
Diethyl carbonate, 99%
C5H10O3