EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Application of a chiral metal-organic framework in enantioselective separation.
PMID 21960280
Abstract
A modular approach for the synthesis of highly ordered porous and chiral auxiliary (Evans auxiliary) decorated metal-organic frameworks is developed. Our synthesis strategy, which uses known porous structures as model materials for incorporation of chirality via linker modification, can provide access to a wide range of porous materials suitable for enantioselective separation and catalysis. Chiral analogues of UMCM-1 have been synthesized and investigated for the enantioseparation of chiral compounds in the liquid phase and first promising results are reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P13800
1-Phenylethanol, 98%
C8H10O
W268518
α-Methylbenzyl alcohol, ≥99%, FCC, FG
C8H10O