EMAIL THIS PAGE TO A FRIEND
Journal of the American Veterinary Medical Association
Severe infestation of bedbugs in a poultry breeder house.
PMID 21961628
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46003
β-Cyfluthrin, PESTANAL®, analytical standard
C22H18Cl2FNO3
33738
Baythroid®, PESTANAL®, analytical standard
C22H18Cl2FNO3