EMAIL THIS PAGE TO A FRIEND
Organic letters
Formation of 1:2 host-guest complexes based on triptycene-derived macrotricycle and paraquat derivatives: anion-π interactions between PF6(-) and bipyridinium rings in the solid state.
PMID 21962056
Abstract
A triptycene-derived macrotricyclic host containing two dibenzo-[30]-crown-10 moieties forms stable 1:2 host-guest complexes with paraquat derivatives in both solution and the solid state, in which anion-π interactions between PF(6)(-) and the bipyridinium rings play an important role. Moreover, it was found that binding and release of the guest molecules in the complexes could be easily controlled by the addition and removal of potassium ions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
332518
Dibenzo-30-crown-10, 98%
C28H40O10