EMAIL THIS PAGE TO A FRIEND
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology
Neuropeptide levels in nasal secretions from patients with and without chronic cough.
PMID 21962097
Abstract
The pathophysiology of upper airway cough syndrome is not well understood. To investigate the levels of neuropeptides found in nasal mucus of subjects with and without the complaint of chronic cough. Preformed nasal mucus from 26 subjects complaining of postnasal drip with chronic cough and 17 without self-reported chronic cough was collected at the time of presentation. The nasal secretions were assayed for 3 neuropeptides-substance P (SP), calcitonin gene-related peptide (CGRP), and Neurokinin A (NKA)-that have been previously linked to chronic cough. Coughing subjects with postnasal drip have significantly higher levels of CGRP and SP than noncoughing subjects with postnasal drip (69.0 ng vs 4.6 ng, P < .001 and 4.5 ng vs 3.4 ng, P = .004). The differences in NKA levels in nasal secretions between coughing and noncoughing subjects did not reach statistical significance (16.3 ng vs 3.4 ng, P = .067). Both CGRP and SP levels are elevated in preformed nasal secretions of coughing subjects when compared with noncoughing subjects. Additional studies are needed to clarify whether CGRP and SP levels in preformed nasal secretions may assist in diagnosis or predicting treatment response in patients presenting with chronic cough.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N4267
Neurokinin A, ≥97% (HPLC)
C50H80N14O14S