EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. A
Assessing the polycyclic aromatic hydrocarbon anisotropic potential with application to the exfoliation energy of graphite.
PMID 21967093
Abstract
In this work we assess a recently published anisotropic potential for polycyclic aromatic hydrocarbon (PAH) molecules (J. Chem. Theory Comput. 2010, 6, 683-695). Comparison to recent high-level symmetry-adapted perturbation theory based on density functional theory (SAPT(DFT)) results for coronene (C(24)H(12)) demonstrate the transferability of the potential while highlighting some limitations with simple point charge descriptions of the electrostatic interaction. The potential is also shown to reproduce second virial coefficients of benzene (C(6)H(6)) with high accuracy, and this is enhanced by using a distributed multipole model for the electrostatic interaction. The graphene dimer interaction energy and the exfoliation energy of graphite have been estimated by extrapolation of PAH interaction energies. The contribution of nonlocal fluctuations in the π electron density in graphite have also been estimated which increases the exfoliation energy by 3.0 meV atom(-1) to 47.6 meV atom(-1), which compares well to recent theoretical and experimental results.