EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Microchip electrophoresis of bacteria using lipid-based liquid crystalline nanoparticles.
PMID 21968347
Abstract
The aggregation and adhesion of bacterial cells is a serious disadvantage for electrophoretic separations of bacteria. In this study, lipid-based liquid crystalline nanoparticles were used as a pseudostationary phase to minimise the bacterial aggregation and adsorption to the inner walls of microchannels. Lactobacillus delbrueckii subsp. bulgaricus, Streptococcus thermophilus and Lactobacillus rhamnosus were selected as analytes and were separated by microchip electrophoresis (MCE) with laser-induced fluorescence (LIF) detection using 4.5 mM tris(hydroxymethyl) aminomethane (TRIS)-4.5 mM boric acid-0.1 mM ethylenediaminetetraacetate (EDTA) (TBE) containing poly(ethylene oxide) (PEO) and lipid-based nanoparticles as the running buffer. The mechanism of lipid-based nanoparticles affecting bacterial adhesion and aggregation was discussed and supported by zeta potential experiments. Under the optimal conditions, the three species of bacteria were identified with patterned peaks. This proposed MCE method using lipid-based nanoparticles as running buffer additives was also used to analyse a real yogurt sample, and valuable bacterial information was obtained by the electropherograms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8894
Dioleoylglycerol, ≥99%, mixture of 1,3- and 1,2-isomers, liquid
C39H72O5