EMAIL THIS PAGE TO A FRIEND
Ocular immunology and inflammation
Lupus relapse presented as frosted branch retinal angiitis: case report.
PMID 21970665
Abstract
To describe a systemic lupus erythematosus (SLE) relapse, which presented as frosted branch retinal angiitis. Clinical case report. A 16-year-old female patient had an SLE relapse that appeared as frosted branch angiitis while being treated with deflazacort and azathioprine. Complete resolution of the vasculitis was achieved with a 3-day course of pulsed intravenous methylprednisolone and oral prednisone 3 weeks later. Even with proper immunosuppressive treatment, SLE can relapse. This case presented as frosted branch retinal angiitis, which is a clinical picture that is rarely described in SLE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0123
Deflazacort, ≥98% (HPLC)
C25H31NO6