EMAIL THIS PAGE TO A FRIEND
The Analyst
Protonating and determining myosmine intactly by association with citrate anion.
PMID 21971428
Abstract
Myosmine can not be separated from nornicotine, nicotine and anabasine intactly by capillary zone electrophoresis (CZE) with a phosphate buffer. Using citrate solution at pH 6.5 as a CZE buffer, myosmine is protonated intactly by H(+), charged positively and then separated from other tobacco alkaloids on the baseline. Its sensitivity is ten times higher than gas chromatography (GC) with a nitrogen-phosphorus detector (NPD). The mechanism for protonating myosmine intactly is discussed and the utility of the new method is testified, too.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8765
Myosmine, ≥98%
C9H10N2