EMAIL THIS PAGE TO A FRIEND
Langmuir : the ACS journal of surfaces and colloids
Effect of submicellar concentrations of conjugated and unconjugated bile salts on the lipid bilayer membrane.
PMID 21973323
Abstract
The interaction of submicellar concentrations of various physiologically important unconjugated [sodium deoxycholate (NaDC), sodium cholate (NaC)] and conjugated [sodium glycodeoxycholate (NaGDC), sodium glycocholate (NaGC), sodium taurodeoxycholate (NaTDC), sodium taurocholate (NaTC)] bile salts with dipalmitoylphosphatidylcholine (DPPC) and dimyristoylphosphatidylcholine (DMPC) small unilamellar vesicles in solid gel (SG) and liquid crystalline (LC) phases was investigated using the excited-state prototropism of 1-naphthol. Steady-state and time-resolved fluorescence of the two excited-state prototropic forms of 1-naphthol indicate that submicellar bile salt concentration induces hydration of the lipid bilayer membrane into the core region. This hydration effect is a general phenomenon of the bile salts studied. The bilayer hydration efficiency of the bile salt follows the order NaDC > NaC > NaGDC > NaTDC > NaGC > NaTC for both DPPC and DMPC vesicles in their SG and LC phases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7132
Sodium glycocholate hydrate, ≥95% (TLC)
C26H42NNaO6 · xH2O
T0875
Sodium taurodeoxycholate hydrate, ≥95% (HPLC)
C26H44NNaO6S · xH2O