EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of carcinogenic risks to humans
Disperse Yellow 3.
PMID 2197457
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
215686
Disperse Yellow 3, Dye content 30 %
C15H15N3O2
11344
Disperse Yellow 3, analytical standard
C15H15N3O2