EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Preparation and characterization of immobilized [A336][MTBA] in PVA-alginate gel beads as novel solid-phase extractants for an efficient recovery of Hg (II) from aqueous solutions.
PMID 21974850
Abstract
The coarse PVA-alginate matrix gel beads entrapping the micro-droplets of the ionic liquid tricaprylylmethylammonium 2-(methylthio) benzoate ([A336][MTBA]) as novel solid-phase extractants were prepared for the removal of mercury (II) from aqueous media. The ionic liquid [A336][MTBA] immobilized PVA-alginate beads (PVA/IL) have been characterized by FTIR, SEM and TGA. The influence of the uptake conditions was investigated including aqueous pH, PVA/IL dosage, the content of [A336][MTBA] and initial Hg (II) concentration; maximum Hg (II) ion adsorption capacity obtained was 49.89 (± 0.11)mgg(-1) at pH 5.8 with adsorptive removal of approximately 99.98%. The selectivity of the PVA/IL beads towards Hg (II), Pb (II) and Cu (II) ions tested was Hg>Pb>Cu. The rate kinetic study was found to follow second-order and the applicability of Langmuir, Freundlich and Tempkin adsorption isotherm model were tested as well. The results of the study showed that PVA/IL beads could be efficiently used as novel extractants for the removal of divalent mercury from aqueous solutions under comparatively easy operation conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69485
Methyltrioctylammonium chloride, ≥97.0% (AT)
C25H54ClN