EMAIL THIS PAGE TO A FRIEND
Environmental health : a global access science source
Heated indoor swimming pools, infants, and the pathogenesis of adolescent idiopathic scoliosis: a neurogenic hypothesis.
PMID 21975145
Abstract
In a case-control study a statistically significant association was recorded between the introduction of infants to heated indoor swimming pools and the development of adolescent idiopathic scoliosis (AIS). In this paper, a neurogenic hypothesis is formulated to explain how toxins produced by chlorine in such pools may act deleteriously on the infant's immature central nervous system, comprising brain and spinal cord, to produce the deformity of AIS. Through vulnerability of the developing central nervous system to circulating toxins, and because of delayed epigenetic effects, the trunk deformity of AIS does not become evident until adolescence. In mature healthy swimmers using such pools, the circulating neurotoxins detected are chloroform, bromodichloromethane, dibromochloromethane, and bromoform. Cyanogen chloride and dichloroacetonitrile have also been detected. In infants, the putative portals of entry to the blood could be dermal, oral, or respiratory; and entry of such circulating small molecules to the brain are via the blood-brain barrier, blood-cerebrospinal fluid barrier, and circumventricular organs. Barrier mechanisms of the developing brain differ from those of adult brain and have been linked to brain development. During the first 6 months of life cerebrospinal fluid contains higher concentrations of specific proteins relative to plasma, attributed to mechanisms continued from fetal brain development rather than immaturity. The hypothesis can be tested. If confirmed, there is potential to prevent some children from developing AIS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
125954
Dichloroacetonitrile, 98%
C2HCl2N
442560
Dichloroacetonitrile, analytical standard
C2HCl2N