EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Visual observation of the mercury-stimulated peroxidase mimetic activity of gold nanoparticles.
PMID 21975577
Abstract
Mercury-stimulated peroxidase mimetic activity of gold nanoparticles was presented, with which a sensitive label-free colorimetric method for Hg(2+) was developed.