EMAIL THIS PAGE TO A FRIEND
Journal of nutrition and metabolism
Distinct effects of calorie restriction and resveratrol on diet-induced obesity and Fatty liver formation.
PMID 21977315
Abstract
The potential of resveratrol to mimic beneficial effects of calorie restriction (CR) was investigated. We compared the effects of both CR (70% of ad libitum energy intake) or resveratrol (2 g/kg or 4 g/kg food) on high-fat diet-induced obesity and fatty liver formation in C57Bl/6J mice, and we examined their effects on calorimetry, metabolic performance, and the expressions of inflammatory genes and SIRT proteins. We found that resveratrol with 4 g/kg dose partially prevented hepatic steatosis and hepatocyte ballooning and induced skeletal muscle SIRT1 and SIRT4 expression while other examined parameter were unaffected by resveratrol. In contrast, CR provided superior protection against diet-induced obesity and fatty liver formation as compared to resveratrol, and the effects were associated with increased physical activity and ameliorated adipose tissue inflammation. CR increased expressions of SIRT3 in metabolically important tissues, suggesting that the beneficial effects of CR are mediated, at least in part, via SIRT3-dependent pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AMPD1
DNase I, Amplification Grade