EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Organo/layered double hydroxide nanohybrids used to remove non ionic pesticides.
PMID 21978582
Abstract
The preparation and characterization of organo/layered double hydroxide nanohybrids with dodecylsulfate and sebacate as interlayer anion were studied in detail. The aim of the modification of the layered double hydroxides (LDHs) was to change the hydrophilic character of the interlayer to hydrophobic to improve the ability of the nanohybrids to adsorb non-ionic pesticides such as alachlor and metolachlor from water. Adsorption tests were conducted on organo/LDHs using variable pH values, contact times and initial pesticide concentrations (adsorption isotherms) in order to identify the optimum conditions for the intended purpose. Adsorbents and adsorption products were characterized several physicochemical techniques. The adsorption test showed that a noticeable increase of the adsorption of the non-ionic herbicides was produced. Based on the results, the organo/LDHs could be good adsorbents to remove alachlor and metolachlor from water. Different organo/LDHs complexes were prepared by a mechanical mixture and by adsorption. The results show that HTSEB-based complex displays controlled release properties that reduce metolachlor leaching in soil columns compared to a technical product and the other formulations. The release was dependent on the nature of the adsorbent used to prepare the complexes. Thus, it can be concluded that organo/LDHs might act as suitable supports for the design of pesticide slow release formulations with the aim of reducing the adverse effects derived from rapid transport losses of the chemical once applied to soils.
Related Materials