EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Hyperoside protects primary rat cortical neurons from neurotoxicity induced by amyloid β-protein via the PI3K/Akt/Bad/Bcl(XL)-regulated mitochondrial apoptotic pathway.
PMID 21978835
Abstract
Amyloid β-protein (Aβ), which is deposited in neurons as neurofibrillary tangles, is known to exert cytotoxic effects by inducing mitochondrial dysfunction. Additionally, the PI3K/Akt-mediated interaction between Bad and Bcl(XL) plays an important role in maintaining mitochondrial integrity. However, the application of therapeutic drugs, especially natural products in Alzheimer's disease therapy via PI3K/Akt/Bad/Bcl(XL)-regulated mitochondrial apoptotic pathway has not aroused extensive attention. In the present study, we investigated the neuroprotective effects of hyperoside, a bioactive flavonoid compound from Hypericum perforatum, on Aβ(25-35)-induced primary cultured cortical neurons, and also examined the potential cellular signaling mechanism for Aβ detoxication. Our results showed that treatment with hyperoside significantly inhibited Aβ(25-35)-induced cytotoxicity and apoptosis by reversing Aβ-induced mitochondrial dysfunction, including mitochondrial membrane potential decrease, reactive oxygen species production, and mitochondrial release of cytochrome c. Further study indicated that hyperoside can activate the PI3K/Akt signaling pathway, resulting in inhibition of the interaction between Bad and Bcl(XL), without effects on the interaction between Bad and Bcl-2. Furthermore, hyperoside inhibited mitochondria-dependent downstream caspase-mediated apoptotic pathway, such as that involving caspase-9, caspase-3, and poly ADP-ribose polymerase (PARP). These results demonstrate that hyperoside can protect Aβ-induced primary cultured cortical neurons via PI3K/Akt/Bad/Bcl(XL)-regulated mitochondrial apoptotic pathway, and they raise the possibility that hyperoside could be developed into a clinically valuable treatment for Alzheimer's disease and other neuronal degenerative diseases associated with mitochondrial dysfunction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00180585
Hyperoside, primary reference standard
C21H20O12
83388
Quercetin 3-D-galactoside, ≥97.0% (HPLC)
C21H20O12