EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Massilia sp. BS-1, a novel violacein-producing bacterium isolated from soil.
PMID 21979084
Abstract
A novel bacterium, Massilia sp. BS-1, producing violacein and deoxyviolacein was isolated from a soil sample collected from Akita Prefecture, Japan. The 16S ribosomal DNA of strain BS-1 displayed 93% homology with its nearest violacein-producing neighbor, Janthinobacterium lividum. Strain BS-1 grew well in a synthetic medium, but required both L-tryptophan and a small amount of L-histidine to produce violacein.
Related Materials