EMAIL THIS PAGE TO A FRIEND
Paediatric anaesthesia
Withdrawal treatment with clonidine after prolonged use of chloral hydrate in a pediatric intensive care patient.
PMID 21981100
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47335-U
Chloral hydrate solution, 1000 μg/mL in acetonitrile, analytical standard
C2H3Cl3O2