EMAIL THIS PAGE TO A FRIEND
Paediatric anaesthesia
Anesthesia for a child suffering from a deletion in the Xp21 loci resulting in Duchenne disease, glycerol kinase deficiency, and congenital adrenal hypoplasia.
PMID 21981102