EMAIL THIS PAGE TO A FRIEND
Allergologia et immunopathologia
Systemic allergic reaction due to intranasal budesonide.
PMID 21982400
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H8885
Hydroxyzine dihydrochloride, ≥98%
C21H27ClN2O2 · 2HCl
H9010
Hydroxyzine pamoate salt, analytical standard
C21H27ClN2O2 · C23H16O6