EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Highly selective and sensitive chromo-fluorogenic detection of the Tetryl explosive using functional silica nanoparticles.
PMID 21983872
Abstract
Silica nanoparticles containing polyamines and thiol groups have been used as probes for the selective detection of Tetryl.
Related Materials