EMAIL THIS PAGE TO A FRIEND
Nihon yakurigaku zasshi. Folia pharmacologica Japonica
[Protection from pulmonary apoptosis: a new therapeutic choice for septic acute lung injury].
PMID 21986062
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O7389
Olprinone hydrochloride, ≥98% (HPLC), solid
C14H10N4O · HCl