EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Simplified synthesis of isotopically labeled 5,5-dimethyl-pyrroline N-oxide.
PMID 21986521
Abstract
5,5-Dimethylpyrroline N-oxide (15N) and 5,5-di(trideuteromethyl)pyrroline N-oxide were synthesized from the respective isotopically labeled 2-nitropropane analogs obtained from the reaction of sodium nitrate with 2-halopropanes. This facile, straightforward process allows synthesizing isotopically labeled DMPO analogs in a 4-step reaction without special equipment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130265
2-Nitropropane, ≥96%
C3H7NO2