EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Synthesis of functionalized 1H-indenes via copper-catalyzed arylative cyclization of arylalkynes with aromatic sulfonyl chlorides.
PMID 21988538
Abstract
A variety of polysubstituted 1H-indenes can be prepared through the copper-catalyzed arylative cyclization of simple arylalkynes with commercially available aromatic sulfonyl chlorides that function as an aryl group donor. The reaction tolerates a broad range of functional groups, including bromide and iodide, nitrile, ketone, and nitro groups. The reaction allowed the synthesis of polycyclic aromatic hydrocarbons, such as a bis(indene), indacene, and fused polyarene derivatives, some of them showing strong fluorescence in solution and the solid state.
Related Materials