EMAIL THIS PAGE TO A FRIEND
Journal of natural products
The thermal and enzymatic taxifolin-alphitonin rearrangement.
PMID 21992235
Abstract
This report describes a detailed investigation of the thermal and enzymatic conversion of taxifolin to alphitonin. Chromatographic separation of the four dihydroquercetin stereoisomers 1-4 in combination with circular dichroism spectroscopy permitted elucidation of the kinetics of this rearrangement and characterization of the different reaction pathways involved. Our findings are corroborated by quantum chemistry calculations that reveal a unique cascade of tautomerization processes leading from taxifolin to alphitonin and also explain the racemization of alphitonin at room temperature. Furthermore, the substrate specificity toward (+)-taxifolin of an enzyme from Eubacterium ramulus catalyzing this intriguing rearrangement is demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78666
Taxifolin, analytical standard
C15H12O7
03890585
Taxifolin, primary reference standard
C15H12O7