EMAIL THIS PAGE TO A FRIEND
Cephalalgia : an international journal of headache
Needling the pain and comforting the brain: acupuncture in the treatment of chronic migraine.
PMID 21994112
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36133
Isazophos, PESTANAL®, analytical standard
C9H17ClN3O3PS