EMAIL THIS PAGE TO A FRIEND
Diagnostic cytopathology
Microfilariae of Wuchereria bancrofti in urine: an uncommon finding.
PMID 21994197
Abstract
Filariasis is a global health problem commonly seen in tropics and subtropics. Microfilariae has been reported in cytological specimens from various sites but is an unusual finding from sediments of centrifuged normally voided urine samples. This highlights the presence of adult worms in the lymphatics and thus helping in prompt decision to start antihelminthic treatment besides providing surgical treatment for chyluria.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8765
Diethylcarbamazine citrate salt, ≥98%
C16H29N3O8