EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
Doping control in horses: housing conditions and oral recycling of flunixin by ingestion of contaminated straw.
PMID 21995754
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0530
Clonixin, ≥98% (HPLC)
C13H11ClN2O2