EMAIL THIS PAGE TO A FRIEND
Allergologia et immunopathologia
Anaphylactic reaction to povidone secondary to drug ingestion in a young child.
PMID 21996435
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0123
Deflazacort, ≥98% (HPLC)
C25H31NO6
H8885
Hydroxyzine dihydrochloride, ≥98%
C21H27ClN2O2 · 2HCl
H9010
Hydroxyzine pamoate salt, analytical standard
C21H27ClN2O2 · C23H16O6