EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
Cuts by caspase-14 control the proteolysis of filaggrin.
PMID 21997417
Abstract
Although mutations in the filaggrin gene (FLG) have been shown to be associated with ichthyosis vulgaris and atopic dermatitis, the function and regulation of filaggrin remain incompletely understood. In this issue, Hoste et al. report that filaggrin is directly cleaved by caspase-14. Acting in concert with other proteases, caspase-14 controls the breakdown of filaggrin to free amino acids and amino acid derivatives that contribute to the hydration and UVB absorption capacity of the stratum corneum. These findings identify a new layer of complexity in the regulation of epidermal barrier function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
859796
4-Imidazoleacrylic acid, 99%
C6H6N2O2