EMAIL THIS PAGE TO A FRIEND
Macromolecular rapid communications
Stimuli-responsive water-soluble fullerene (C60) polymeric systems.
PMID 21997830
Abstract
This review documents the advances in stimuli-responsive water-soluble fullerene (C(60)) polymeric systems. Stimuli-responsive polymers, when grafted onto C(60) impart "smart" and "responsive" characteristics, and these novel materials adopt various morphologies when subjected to external stimuli, such as pH, temperature, and salt. Various synthetic approaches for producing C(60)-polymers are outlined and discussed. The responsive behavior, water solubility, and self-assembly characteristics of these C(60)-polymers make them attractive for applications such as drug delivery, temperature sensors, and personal care.
Related Materials