EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A convergent approach to polycyclic aromatic hydrocarbons.
PMID 21998824
Abstract
A new concise route to Polycyclic Aromatic Hydrocarbons (PAHs) through radical addition and cyclisation of xanthates is described.