EMAIL THIS PAGE TO A FRIEND
FEBS letters
Serine protease inhibitor mediated peptide bond re-synthesis in diverse protein molecules.
PMID 22001203
Abstract
Protease inhibitors have been extensively used in research to prevent unwanted degradation of proteins during purification and analysis. Here, we report a remarkable discovery of protease inhibitor mediated reformation of peptide bonds by the serine protease inhibitor, PMSF in a diverse set of proteolyzed molecules. Interestingly, the religation reaction in the presence of PMSF occurs in a very short time period and with very high yields of the religated product. We also investigate the plausible mechanism of such a reaction and demonstrate through biochemical studies and X-ray crystallography that proximity of reacting termini is essential for the feasibility of this reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93482
Phenylmethanesulfonyl fluoride solution, ~0.1 M in ethanol (T)