EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Site-specific incorporation of arginine analogs into proteins using arginyl-tRNA synthetase.
PMID 22001918
Abstract
Arginine analogs were incorporated site-specifically into proteins using an in vitro translation system. In this system, mRNAs containing a CGGG codon were translated by an aminoacyl-tRNA(CCCG), which was charged with arginine analogs using yeast arginyl-tRNA synthetase. N(G)-monomethyl-L-arginine, L-citrulline and L-homoarginine were incorporated successfully into proteins using this method. The influence of arginine monomethylation in histone H3 on the acetylation of lysine residues by histone acetyltransferase hGCN5 was investigated, and the results demonstrated that K9 acetylation was suppressed by the methylation of R8 and R17 but not by R26 methylation. K18 acetylation was not affected by the methylation of R8, R17 and R26. This site-specific modification strategy provides a way to explore the roles of post-translational modifications in the absence of heterogeneity due to other modifications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1007
L-Homoarginine hydrochloride, unnatural arginine analog
C7H16N4O2 · HCl