EMAIL THIS PAGE TO A FRIEND
Journal of food protection
Distribution of T-2 and HT-2 toxins in milling fractions of durum wheat.
PMID 22004818
Abstract
The effect of processing on mycotoxin content in milling fractions has been investigated in 10 samples of durum wheat contaminated with T-2 and HT-2 toxins at levels ranging from 97 to 5,954 μg/kg (sum of T-2 and HT-2 toxins). Either naturally contaminated samples or samples artificially inoculated with Fusarium sporotrichioides under field conditions were used. A method based on liquid chromatography-tandem mass spectrometry coupled with immunoaffinity column cleanup was validated in-house for the simultaneous analysis of both toxins in a variety of matrices, including uncleaned wheat, cleaned wheat, screenings, bran, red dog, fine middlings, and semolina. Mean recoveries from samples spiked with T-2 and HT-2 toxins at levels of 100 μg/kg ranged from 85 to 107%, with relative standard deviations (RSDs) lower than 14%. The milling process led to an increase of T-2 and HT-2 toxin contents up to 13- and 5-fold in screenings and bran, respectively, compared with occurrence in the uncleaned wheat; however, an overall reduction of T-2 and HT-2 toxins by 54% (RSD, 20%) and 89% (RSD, 3%) was observed in cleaned wheat and in semolina, respectively.