EMAIL THIS PAGE TO A FRIEND
Archives of dermatology
Localized skin necrosis of steroid-induced striae distensae: an unusual complication of bevacizumab and irinotecan therapy.
PMID 22006149
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3781
6α-Methylprednisolone 21-hemisuccinate sodium salt, lyophilized powder
C26H33NaO8